EMAIL THIS PAGE TO A FRIEND
Cancer chemotherapy and pharmacology
Effects of famotidine or an antacid preparation on the pharmacokinetics of nilotinib in healthy volunteers.
PMID 23070146
Abstract
This study evaluated the effects of either famotidine or antacid on the pharmacokinetics of nilotinib in healthy subjects, with the specific focus to explore different dosing separation schemes leading to a minimized drug-drug interaction. Fifty-two subjects were randomized to receive the following treatments in a crossover manner: (A) single oral nilotinib 400 mg alone; (B) famotidine 20 mg twice a day for 3 days, followed by a single administration of nilotinib 400 mg and famotidine 20 mg on Day 4, where famotidine was given 2 h after nilotinib; (C) single oral nilotinib 400 mg and antacid suspension 20 mL, where antacid was given 2 h before nilotinib; (D) single oral nilotinib 400 mg and antacid suspension 20 mL, where antacid was given 2 h after nilotinib. Comparing Treatment B to Treatment A, the geometric mean ratios of nilotinib C(max), AUC(0-tlast), and AUC(0-inf) were 0.966, 0.984, and 0.911, respectively (90% confidence intervals (CIs), 0.875-1.066, 0.905-1.069, and 0.798-1.039, respectively). Nilotinib pharmacokinetic parameters following Treatment C or Treatment D were similar to those after Treatment A; the corresponding 90% CIs of the geometric mean ratios of C(max), AUC(0-tlast), and AUC(0-inf) all fell within the bioequivalence range of 0.8-1.25. Neither famotidine nor antacid significantly affected nilotinib pharmacokinetics. When concurrent use of an H2 blocker or an antacid is necessary, the H2 blocker may be administered 10 h before and 2 h after nilotinib dose, or the antacid may be administered 2 h before or 2 h after nilotinib dose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1055
Famotidine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H15N7O2S3