EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Liposome-mediated delivery of iminosugars enhances efficacy against dengue virus in vivo.
PMID 23070155
Abstract
A key challenge faced by promising antiviral drugs, such as iminosugars, is in vivo delivery to achieve effective levels of drug without toxicity. Four iminosugars, all deoxynojirimycin (DNJ) derivatives-N-butyl DNJ (NB-DNJ), N-nonyl DNJ, N-(9-methoxynonyl) DNJ, and N-(6'-[4″-azido-2″-nitrophenylamino]hexyl)-1-DNJ (NAP-DNJ)-potently inhibited both the percentage of cells infected with dengue virus and release of infectious virus from primary human monocyte-derived macrophages, demonstrating their efficacy in primary cells. In a lethal antibody-dependent enhancement mouse model of dengue pathogenesis, free NB-DNJ significantly enhanced survival and lowered viral load in organs and serum. Liposome-mediated delivery of NB-DNJ, in comparison with free NB-DNJ, resulted in a 3-log(10) reduction in the dose of drug sufficient to enhance animal survival. The optimizing of the effective dose in this way could liberate the therapeutic potential of many cytotoxic antivirals against both dengue virus and a wide array of other viruses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9305
1-Deoxynojirimycin hydrochloride
C6H13NO4 · HCl