EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Investigations regarding the utility of prodigiosenes to treat leukemia.
PMID 23070266
Abstract
Prodigiosenes, possessing a 4-methoxypyrrolyldipyrrin skeleton, are known for their anti-cancer activity. Structural modification of the C-ring resulted in a series of prodigiosenes that displayed promising activity against leukemia cell lines during in vitro analysis against the NCI 60 cancer cell line panel. Further in vivo studies of these compounds using the zebrafish model showed persistence of anti-leukemia properties in human K562 chronic myelogenous leukemia cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
537594
tert-Butyl acetoacetate, reagent grade, 98%
C8H14O3