PloS one
Oroxylin-A rescues LPS-induced acute lung injury via regulation of NF-κB signaling pathway in rodents.
PMID 23071799
Successful drug treatment for sepsis-related acute lung injury (ALI) remains a major clinical problem. This study was designed to assess the beneficial effects of post-treatment of oroxylin A (OroA), a flavonoid, in ameliorating lipopolysaccharides (LPS)-induced lung inflammation and fatality. Rats were injected with LPS (10 mg/kg, iv) to induce ALI, and OroA was given (15 mg/kg, iv) 1 hr or 6 hrs after LPS challenge. Twenty four hrs after LPS challenge, biochemical changes in the blood and lung tissues, and morphological/histological alterations in the lung associated with inflammation and injury were examined. Therapeutic effect of OroA was assessed by measuring the survival rate in endotoxemic mice. LPS (10 mg/kg, iv) significantly altered WBC counts, elevated plasma tumor necrosis factor (TNF)-α and nitric oxide (NO), increased pulmonary edema, thickened alveolar septa, and decreased survival rate. These changes were ameliorated by OroA (15 mg/kg, iv) administered 1 hr or 6 hrs after LPS challenge. This post-treatment also significantly attenuated LPS-induced activation of nuclear factor-κB (NF-κB) and the release of high mobility group box 1 (HMGB1) in lung tissues. Furthermore, post-treatment with OroA (60 mg/kg, ip) administered 1 hr or 6 hrs after LPS challenge in mice significantly increased survival rate. OroA administered after induction of ALI by LPS significantly prevent and revere lung tissues injuries with increased survival rate. Positive post-treatment effects of OroA suggest that OroA is a potentially useful candidate for managing lung inflammation in LPS-induced endotoxemia and septic shock.