EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutical compounding
Medical therapy for acquired urinary incontinence in dogs.
PMID 23072196
Abstract
The objective of this case report was to review acquired canine urethral incontinence in a case of a 2-year-old beagle that presented for inappropriate urination. Commercially available medications, as well as compounded medical therapeutic options, were reviewed. Diagnostic results revealed cystitis, which responded to the prescribed antibiotics. The incontinence issue remained. Compounded diethylstilbestrol was prescribed and dispensed to achieve successful clinical results. The role of the compounding pharmacist was vital to achieve appropriate dosing of a drug no longer commercially available.