EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Integrated delignification and simultaneous saccharification and fermentation of hard wood by a white-rot fungus, Phlebia sp. MG-60.
PMID 23073100
Abstract
We propose a new process of unified aerobic delignification and anaerobic saccharification and fermentation of wood by a single microorganism, the white-rot fungus Phlebia sp. MG-60. This fungus is able to selectively degrade lignin under aerobic solid state fermentation conditions, and to produce ethanol directly from delignified oak wood under semi-aerobic liquid culture conditions. After 56 d aerobic incubation, 40.7% of initial lignin and negligible glucan were degraded. Then under semi-aerobic conditions without the addition of cellulase, 43.9% of theoretical maximum ethanol was produced after 20 d. Changing from aerobic conditions (biological delignification pretreatment) to semi-aerobic conditions (saccharification and fermentation) enabled the fermentation of wood by solely biological processes. This is the first report of ethanol production from woody biomass using a single microorganism without addition of chemicals or enzymes.
Related Materials