EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Simultaneous production of bio-ethanol and bleached pulp from red algae.
PMID 23073109
Abstract
The red algae, Gelidium corneum, was used to produce bleached pulp for papermaking and ethanol. Aqueous extracts obtained at 100-140 °C were subjected to saccharification, purification, fermentation, and distillation to produce ethanol. The solid remnants were bleached with chlorine dioxide and peroxide to make pulp. In the extraction process, sulfuric acid and sodium thiosulfate were added to increase the extract yield and to improve de-polymerization of the extracts, as well as to generate high-quality pulp. An extraction process incorporating 5% sodium thiosulfate by dry weight of the algae provided optimal production conditions for the production of both strong pulp and a high ethanol yield. These results suggest that it might be possible to utilize algae instead of trees and starch for pulp and ethanol production, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53407
5-(Hydroxymethyl)furfural, analytical standard
C6H6O3