EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Microalgae cultivation in wastewater: nutrient removal from anaerobic membrane bioreactor effluent.
PMID 23073115
Abstract
This study investigated the removal of nitrogen and phosphorus from the effluent of a submerged anaerobic membrane bioreactor (SAnMBR) by means of a lab-scale photobioreactor in which algae biomass was cultured in a semi-continuous mode for a period of 42 days. Solids retention time was 2 days and a stable pH value in the system was maintained by adding CO(2). Nitrogen and phosphorus concentrations in the SAnMBR effluent fluctuated according to the operating performance of the bioreactor and the properties of its actual wastewater load. Despite these variations, the anaerobic effluent proved to be a suitable growth medium for microalgae (mean biomass productivity was 234 mg l(-1)d(-1)), achieving a nutrient removal efficiency of 67.2% for ammonium (NH(4)(+)-N) and 97.8% for phosphate (PO(4)(-3)-P). When conditions were optimum, excellent water quality with very low ammonium and phosphate concentrations was obtained.
Related Materials