EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Solid state grinding as a tool to aid enantiomeric resolution by cocrystallisation.
PMID 23073186
Abstract
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single enantiomer of tartaric acid.