EMAIL THIS PAGE TO A FRIEND
Harvard health letter
Ask the doctor. I use a lot of canola oil in cooking, because it's heart-healthy. My husband has prostate cancer, and I just heard that canola oil might make it worse. Is that really true and should I stop using it for cooking?
PMID 23074721