EMAIL THIS PAGE TO A FRIEND
Pain medicine (Malden, Mass.)
Flupirtine enhances the anti-hyperalgesic effects of morphine in a rat model of prostate bone metastasis.
PMID 23078152
Abstract
Current treatments for cancer pain are often inadequate, particularly when metastasis to bone is involved. The addition to the treatment regimen of another drug that has a complementary analgesic effect may increase the overall analgesia without the necessity to increase doses, thus avoiding dose-related side effects. This project investigated the synergistic effect of the addition of the potassium channel (KCNQ2-3) modulator flupirtine to morphine treatment in a rat model of prostate cancer-induced bone pain. Syngeneic prostate cancer cells were injected into the right tibia of male Wistar rats under anesthesia. This led to expanding tumor within the bone in 2 weeks, together with the concurrent development of hyperalgesia to noxious heat. Paw withdrawal thresholds from noxious heat were measured before and after the maximum non-sedating doses of morphine and flupirtine given alone and in combinations. Dose-response curves for morphine (0.13-5.0 mg/kg ip) and flupirtine (1.25-10.0 mg/kg ip) given alone and in fixed-dose combinations were plotted and subjected to an isobolographic analysis. Both morphine (ED₅₀ = 0.74 mg/kg) and flupirtine (ED₅₀ = 3.32 mg/kg) caused dose-related anti-hyperalgesia at doses that did not cause sedation. Isobolographic analysis revealed that there was a synergistic interaction between flupirtine and morphine. Addition of flupirtine to morphine treatment improved morphine anti-hyperalgesia, and resulted in the reversal of cancer-induced heat hyperalgesia. These results suggest that flupirtine in combination with morphine may be useful clinically to provide better analgesia at lower morphine doses in the management of pain caused by tumors growing in bone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8927
Flupirtine maleate salt, ≥98% (HPLC)
C15H17FN4O2 · C4H4O4