EMAIL THIS PAGE TO A FRIEND
The Medical journal of Malaysia
Citrobacter koseri bacteraemia complicated by paraspinal abscess and spondylodiscitis--a case report.
PMID 23082432
Abstract
Paraspinal abscess and spondylodiscitis due to Citrobacter koseri is a very rare condition. We report a remarkable case of Citrobacter koseri bacteraemia complicated by paraspinal abscess and spondylodiscitis in a patient who has successfully been treated in our hospital. Our patient demonstrates one of the common challenges in the practice of infectious disease medicine, wherein an innocuous presentation may and often underlie a serious infection. This case report elucidates to us that the diagnosis of a paraspinal abscess and spondylodiscitis requires a high index of suspicion in at risk patient presenting with compatible signs and symptoms.