EMAIL THIS PAGE TO A FRIEND
Malaria journal
Therapeutic efficacy of artemether-lumefantrine for Plasmodium vivax infections in a prospective study in Guyana.
PMID 23083017
Abstract
In Guyana, chloroquine + primaquine is used for the treatment of vivax malaria. A worldwide increase of chloroquine resistance in Plasmodium vivax led to questioning of the current malaria treatment guidelines. A therapeutic efficacy study was conducted using artemether-lumefantrine + primaquine against P. vivax to evaluate a treatment alternative for chloroquine. From 2009 to 2010, a non-controlled study in two hospitals in Guyana was conducted. A total 61 patients with P. vivax infection were treated with artemether-lumefantrine as a six-dose regimen twice a day for three days with additional 0.25 mg/kg/d primaquine at day 0 for 14 days. Clinical and parasitological parameters were followed on days 0,1,2,3,7,14 and 28 in agreement with WHO guidelines. Plasmodium vivax DNA from eight patients was analysed for pvmdr1, molecular marker of resistance. Artemether-lumefantrine cleared 100% of parasites on day 1, but two patients (3%) had recurrence of parasites on day 28, suggesting relapse. No pvmdr1 Y976F polymorphism was detected. The treatment regimen was well tolerated. In Guyana, artemether-lumefantrine represents an adequate treatment option against P. vivax when combined with primaquine. Availability of this alternative will be of great importance in case of emerging chloroquine resistance against P. vivax.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160393
Primaquine bisphosphate, 98%
C15H21N3O · 2H3PO4