EMAIL THIS PAGE TO A FRIEND
Neuron
A subtype-specific function for the extracellular domain of neuroligin 1 in hippocampal LTP.
PMID 23083734
Abstract
At neuronal excitatory synapses, two major subtypes of the synaptic adhesion molecule neuroligin are present. These subtypes, neuroligin 1 and neuroligin 3, have roles in synaptogenesis and synaptic maintenance that appear largely overlapping. In this study, we combine electrophysiology with molecular deletion and replacement of these proteins to identify similarities and differences between these subtypes. In doing so, we identify a subtype-specific role in LTP for neuroligin 1 in young CA1, which persists into adulthood in the dentate gyrus. As neuroligin 3 showed no requirement for LTP, we constructed chimeric proteins of the two excitatory neuroligin subtypes to identify the molecular determinants particular to the unique function of neuroligin 1. Using in vivo molecular replacement experiments, we find that these unique functions depend on a region in its extracellular domain containing the B site splice insertion previously shown to determine specificity of neurexin binding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6816
(±)-AMPA, solid
C7H10N2O4
A0326
(S)-AMPA, ≥98%
C7H10N2O4
N183
NBQX disodium salt hydrate, ≥98% (HPLC)
C12H6N4Na2O6S · xH2O
N171
NBQX hydrate, powder, ≥98% (HPLC)
C12H8N4O6S · xH2O