EMAIL THIS PAGE TO A FRIEND
Journal of the American College of Cardiology
Not all beta-blockers are equal in the management of long QT syndrome types 1 and 2: higher recurrence of events under metoprolol.
PMID 23083782
Abstract
The purpose of this study was to compare the efficacy of beta-blockers in congenital long QT syndrome (LQTS). Beta-blockers are the mainstay in managing LQTS. Studies comparing the efficacy of commonly used beta-blockers are lacking, and clinicians generally assume they are equally effective. Electrocardiographic and clinical parameters of 382 LQT1/LQT2 patients initiated on propranolol (n = 134), metoprolol (n = 147), and nadolol (n = 101) were analyzed, excluding patients <1 year of age at beta-blocker initiation. Symptoms before therapy and the first breakthrough cardiac events (BCEs) were documented. Patients (56% female, 27% symptomatic, heart rate 76 ± 16 beats/min, QTc 472 ± 46 ms) were started on beta-blocker therapy at a median age of 14 years (interquartile range: 8 to 32 years). The QTc shortening with propranolol was significantly greater than with other beta-blockers in the total cohort and in the subset with QTc >480 ms. None of the asymptomatic patients had BCEs. Among symptomatic patients (n = 101), 15 had BCEs (all syncopes). The QTc shortening was significantly less pronounced among patients with BCEs. There was a greater risk of BCEs for symptomatic patients initiated on metoprolol compared to users of the other 2 beta-blockers combined, after adjustment for genotype (odds ratio: 3.95, 95% confidence interval: 1.2 to 13.1, p = 0.025). Kaplan-Meier analysis showed a significantly lower event-free survival for symptomatic patients receiving metoprolol compared to propranolol/nadolol. Propranolol has a significantly better QTc shortening effect compared to metoprolol and nadolol, especially in patients with prolonged QTc. Propranolol and nadolol are equally effective, whereas symptomatic patients started on metoprolol are at a significantly higher risk for BCEs. Metoprolol should not be used for symptomatic LQT1 and LQT2 patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1892
Nadolol, analytical standard
C17H27NO4