EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Toxic effects of microcystin-LR on the reproductive system of male Rana nigromaculata in vitro.
PMID 23084047
Abstract
This study aims to demonstrate that microcystin-LR (MC-LR) has toxic effects on the reproductive system of male Rana nigromaculata in vitro. R. nigromaculata were treated with 0, 0.1, 1, 10, and 100 nmol/L of MC-LR for 6 h. Results show that exposure to 1 nmol/L to 100 nmol/L of MC-LR decreased sperm motility and number of sperm cells and increased the sperm abnormality rate, whose values were significantly different from those of the control (P<0.01). Moreover, the same dosage of MC-LR increased reactive oxygen species production and malondialdehyde content. At the same time, antioxidant enzyme (catalase and glutathione S-transferase) activity and glutathione reduced content rapidly increased, whereas antioxidant enzyme superoxide dismutase activity significantly decreased. These results imply that the defense system of the testes quickly responds to oxidative stress. Ultrastructural observation shows distention of the mitochondria, endoplasmic reticulum, and Golgi apparatus and changes in the mitochondrial matrix color, cristae number, and morphology. Moreover, using real-time PCR, increased relative expressions of P450 aromatase and SF-1 genes were observed. The results demonstrate for the first time that MC-LR could induce toxicity in the male reproductive system of R. nigromaculata. The findings in this research will provide more insights into the relationships between aquatic microcystins and amphibians.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33893
Microcystin-LR solution, 10 μg/mL in methanol, analytical standard
C49H74N10O12