EMAIL THIS PAGE TO A FRIEND
Journal of pediatric surgery
Re: Novel use of glycopyrrolate (Robinul) in the treatment of anastomotic leak after repair of esophageal atresia and trachea-esophageal fistula.
PMID 23084219