EMAIL THIS PAGE TO A FRIEND
Structure (London, England : 1993)
Crystal structure of a Ba(2+)-bound gating ring reveals elementary steps in RCK domain activation.
PMID 23085076
Abstract
RCK domains control activity of a variety of K(+) channels and transporters through binding of cytoplasmic ligands. To gain insight toward mechanisms of RCK domain activation, we solved the structure of the RCK domain from the Ca(2+)-gated K(+) channel, MthK, bound with Ba(2+), at 3.1 Å resolution. The Ba(2+)-bound RCK domain was assembled as an octameric gating ring, as observed in structures of the full-length MthK channel, and shows Ba(2+) bound at several positions. One of the Ba(2+) sites, termed C1, overlaps with a known Ca(2+)-activation site, determined by residues D184 and E210. Functionally, Ba(2+) can activate reconstituted MthK channels as observed in electrophysiological recordings, whereas Mg(2+) (up to 100 mM) was ineffective. Ba(2+) activation was abolished by the mutation D184N, suggesting that Ba(2+) activates primarily through the C1 site. Our results suggest a working hypothesis for a sequence of ligand-dependent conformational changes that may underlie RCK domain activation and channel gating.