EMAIL THIS PAGE TO A FRIEND
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
National marrow donor program session: donor issues.
PMID 23085601
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001173 Filgrastim, European Pharmacopoeia (EP) Reference Standard