EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and photophysical study of 2'-deoxyuridines labeled with fluorene derivatives.
PMID 23085656
Abstract
We examined microenvironment-sensitive fluorescent 2'-deoxyuridines labeled with fluorene derivatives that exhibited solvent-dependent photophysical properties. The high sensitivity of the fluorescence shift and the nucleoside intensity dependence on solvent polarity provided information useful for estimating the polarity of the environment surrounding the fluorescent nucleoside.