EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Asymmetric trifluoromethylation of aromatic aldehydes by cooperative catalysis with (IPr)CuF and quinidine-derived quaternary ammonium salt.
PMID 23085678
Abstract
A general enantioselective trifluoromethylation of aldehydes has been developed using (IPr)CuF and quinidine-derived quaternary ammonium salt as the cooperative catalyst. Thus, a wide range of aromatic aldehydes have been converted to the corresponding products in up to 92% yield and 81% ee at 2 mol% of catalyst loading. The greatly enhanced activity and enantioselectivity result from the initiative generation of active [(IPr)CuCF(3)] as well as additional coordination activation of other copper species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217905
Copper(II) fluoride, 98%
CuF2