EMAIL THIS PAGE TO A FRIEND
Blood
The clinical significance of NOTCH1 and SF3B1 mutations in the UK LRF CLL4 trial.
PMID 23086750
Abstract
NOTCH1 and SF3B1 mutations have been previously reported to have prognostic significance in chronic lymphocytic leukemia but to date they have not been validated in a prospective, controlled clinical trial. We have assessed the impact of these mutations in a cohort of 494 patients treated within the randomized phase 3 United Kingdom Leukaemia Research Fund Chronic Lymphocytic Leukemia 4 (UK LRF CCL4) trial that compared chlorambucil and fludarabine with and without cyclophosphamide in previously untreated patients. We investigated the relationship of mutations in NOTCH1 (exon 34) and SF3B1 (exon 14-16) to treatment response, survival and a panel of established biologic variables. NOTCH1 and SF3B1 mutations were found in 10% and17% of patients, respectively. NOTCH1 mutations correlated with unmutated IGHV genes, trisomy 12, high CD38/ ZAP-70 expression and were associated with reduced overall (median 54.8 vs 74.6 months, P = .02) and progression-free (median 22.0 vs 26.4 months, P = .02) survival. SF3B1 mutations were significantly associated with high CD38 expression and with shorter overall survival (median 54.3 vs 79.0 months, P < .001). Furthermore, multivariate analysis, including baseline clinical variables, treatment, and adverse prognostic factors demonstrated that although TP53 alterations remained the most informative marker of dismal survival in this cohort, NOTCH1 (HR 1.58, P = .03) and SF3B1 (HR 1.52, P = .01) mutations have added independent prognostic value.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0253
Chlorambucil
C14H19Cl2NO2