EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Preparation of gold nanocluster bioconjugates for electron microscopy.
PMID 23086882
Abstract
In this chapter, we describe types of gold nanoparticle-biomolecule conjugates and their use in electron microscopy. Included are two detailed protocols for labeling an IgG antibody with gold monolayer protected clusters. The first approach is a direct bonding approach that utilizes the ligand place exchange reaction. The second approach describes NHS-EDC coupling of Au(144)(pMBA)(60) with IgG. Also included are various characterization techniques for determining labeling efficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
662534
4-Mercaptobenzoic acid, technical grade, 90%
C7H6O2S
706329
4-Mercaptobenzoic acid, 99%
C7H6O2S