EMAIL THIS PAGE TO A FRIEND
Organic letters
Stereoselective synthesis of conjugated trienols from allylic alcohols and 1-iodo-1,3-dienes.
PMID 23088349
Abstract
The stereoselective synthesis of conjugated trienes has been achieved from allylic alcohols and 1-iodo-1,3-dienes using Pd(OAc)(2)/AgOAc.