EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Prussian blue nanoparticles operate as a new generation of photothermal ablation agents for cancer therapy.
PMID 23090583
Abstract
Herein, prussian blue nanoparticles, an ancient dye, were explored as a new generation of near-infrared laser-driven photothermal ablation agents for cancer therapy alternative to traditional agents due to their good photothermal efficiency and high photothermal stability but low cost and particularly clinically approved biosafety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234125
Iron(III) ferrocyanide
C18Fe7N18