EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Thermal and photochemical reactions of NO2 on chromium(III) oxide surfaces at atmospheric pressure.
PMID 23090708
Abstract
While many studies of heterogeneous chemistry on Cr(2)O(3) surfaces have focused on its catalytic activity, less is known about chemistry on this surface under atmospheric conditions. We report here studies of the thermal and photochemical reactions of NO(2) on Cr(2)O(3) at one atm in air. In order to follow surface species, the interaction of 16-120 ppm NO(2) with a 15 nm Cr(2)O(3) thin film deposited on a germanium crystal was monitored in a flow system using attenuated total reflectance (ATR) coupled to a Fourier transform infrared (FTIR) spectrometer. Gas phase products were monitored in the effluent of an ~285 ppm NO(2)-air mixture that had passed over Cr(2)O(3) powder in a flow system. A chemiluminescence NO(y) analyzer, a photometric O(3) analyzer and a long-path FTIR spectrometer were used to probe the gaseous products. In the absence of added water vapor, NO(2) formed nitrate (NO(3)(-)) ions coordinated to Cr(3+). These surface coordinated NO(3)(-) were reversibly solvated by water under humid conditions. In both dry and humid cases, nitrate ions decreased during irradiation of the surface at 302 nm, and NO and NO(2) were generated in the gas phase. Under dry conditions, NO was the major gaseous product while NO(2) was the dominant species in the presence of water vapor. Heating of the surface after exposure to NO(2) led to the generation of both NO(2) and NO under dry conditions, but only NO(2) in the presence of water vapor. Elemental chromium incorporated into metal alloys such as stainless steel is readily oxidized in contact with ambient air, forming a chromium-rich metal oxide surface layer. The results of these studies suggest that active photo- and thermal chemistry will occur when boundary layer materials containing chromium(III) or chromium oxide such as stainless steel, roofs, automobile bumpers etc. are exposed to NO(2) under tropospheric conditions.