EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Acute Chagas disease in a returning traveler.
PMID 23091192
Abstract
Acute Chagas disease is rarely recognized, and the risk for acquiring the disease is undefined in travelers to Central America. We describe a case of acute Chagas disease in a traveler to Costa Rica and highlight the need for increased awareness of this infection in travelers to Chagas-endemic areas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3415
Nifurtimox, ≥98% (HPLC)
C10H13N3O5S