EMAIL THIS PAGE TO A FRIEND
The Korean journal of pain
Effects of Ethyl Pyruvate on Allodynia, TNF-α Expression, and Apoptosis in the Dorsal Root Ganglion after Spinal Nerve Ligation Injury.
PMID 23091681
Abstract
It has been demonstrated that the expression of tumor necrosis factor-α (TNF-α) and apoptotic cell death in the dorsal root ganglion (DRG) following spinal nerve constriction injury play a role in the initiation and continuation of hyperalgesia and allodynia. The present study was designed to investigate the effects of ethyl pyruvate (EP) on mechanical and cold allodynia, TNF-α expression, and apoptosis in DRG after spinal nerve ligation injury. Rats were divided into 3 groups: control, pre-EP, and post-EP. EP (50 mg/kg) was intraperitoneally injected 30 minutes before (pre-EP) or after (post-EP) surgery. Behavioral tests to determine mechanical and cold allodynia were conducted before surgery and 4 and 7 days after surgery. Seven days after surgery, TNF-α protein levels in DRG were evaluated by enzyme-linked immunosorbent assay, and DRG apoptosis was determined by immunohistochemical detection of activated caspase-3. Treatment with EP significantly reduced mechanical and cold allodynia following spinal nerve ligation injury. TNF-α protein levels in the pre-EP (4.7 ± 1.2 pg/200 µg; P < 0.001) and post-EP (6.4 ± 1.8 pg/200 µg; P < 0.001) groups were 2-3 times lower than the control group (14.4 ± 1.2 pg/200 µg). The percentages of neurons and satellite cells that co-localized with caspase-3 were also significantly lower in the pre-EP and post-EP groups than the control group. These results demonstrate that EP has a strong anti-allodynic effect that acts through the inhibition of TNF-α expression and apoptosis in DRG after spinal nerve ligation injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse