EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
Pharmacokinetics of ginkgolide B injection in beagle dogs.
PMID 23093483
Abstract
A liquid chromatography-mass spectrometry method was developed, validated, and applied to the pharmacokinetic study with doses of 0.68, 2.73 and 10.92 mg/kg of ginkgolide B in beagle dogs after intravenous infusion.An aliquot of blood samples were -collected, separated and quantitatively analyzed by liquid chromatography-mass spectrometry method with mobile phase of acetonitrile-0.02% ammonia solution (33:67, v/v) at a flow rate of 0.8 mL/min on the UltimateTM XB-C18 column (5 μm, 4.6×150 mm).The method was sensitive, accurate and convenient, and can be used for the determination of ginkgolide B in beagle dogs. The Cmax and AUC0-∞ of GB increased with dose escalation, but ANOVA analyses showed that no significant difference was observed in other pharmacokinetic parameters between different doses.An LC/MS method was developed with good sensitivity, reproducibility and specificity. In the pharmacokinetic study of GB in beagle dogs, linear pharmacokinetics was found at doses from 0.62 to 10.92 mg/kg after a single-dose intravenous infusion. Gender differences were not observed in the pharmacokinetics of GB.
Related Materials