EMAIL THIS PAGE TO A FRIEND
BMC research notes
Functional annotation and ENU.
PMID 23095518
Abstract
Functional annotation of every gene in the mouse genome is a herculean task that requires a multifaceted approach. Many large-scale initiatives are contributing to this undertaking. The International Knockout Mouse Consortium (IKMC) plans to mutate every protein-coding gene, using a combination of gene trapping and gene targeting in embryonic stem cells. Many other groups are performing using the chemical mutagen ethylnitrosourea (ENU) or transpon-based systems to induce mutations, screening offspring for phenovariants and identifying the causative mutations. A recent paper in BMC Research Notes by Arnold et al. presents data from an ENU-based mutagenesis project that provides not only some of the first phenotype-genotype information for a large number of genes, but also a trove of information, all publicly available, that demonstrates the specificity and efficiency of ENU mutagenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3385
N-Nitroso-N-ethylurea, ISOPAC®
C3H7N3O2
N8509
N-Nitroso-N-ethylurea, Bulk package
C3H7N3O2