EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Aqueous synthesis of 1-H-2-substituted benzimidazoles via transition-metal-free intramolecular amination of aryl iodides.
PMID 23095894
Abstract
A straightforward method has been developed for the synthesis of the benzimidazole ring system through a carbon-nitrogen cross-coupling reaction. In the presence of 2.0 equiv. of K(2)CO(3) in water at 100 °C for 30 h, the intramolecular cyclization of N-(2-iodoaryl)benzamidine provides benzimidazole derivatives in moderate to high yields. Remarkably, the procedure occurs exclusively in water and doesn’t require the use of any additional reagent/catalyst, rendering the methodology highly valuable from both environmental and economical points of view.