EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Peptidic macrocyclization via palladium-catalyzed chemoselective indole C-2 arylation.
PMID 23096995
Abstract
A highly efficient macrocyclization reaction has been developed via the palladium-catalyzed C-H arylation of the side-chains of tryptophan with halophenyl-containing amino acids. This method allows for direct access to 15- to 25-membered biaryl macrocycles in 40-75% yield, at moderate concentration, with C-H arylation proceeding exclusively at the C-2 position of the tryptophan indole.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48157 Indole solution, 2000 μg/mL in methanol, certified reference material