EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Development of Zn-ProPhenol-catalyzed asymmetric alkyne addition: synthesis of chiral propargylic alcohols.
PMID 23097281
Abstract
The development of a general and practical zinc-catalyzed enantioselective alkyne addition methodology is reported. The commercially available ProPhenol ligand (1) has facilitated the addition of a wide range of zinc alkynylides to aryl, aliphatic, and α,β-unsaturated aldehydes in high yield and enantioselectivity. New insights into the mechanism of this reaction have resulted in a significant reduction in reagent stoichiometry, enabling the use of precious alkynes and avoiding the use of excess dimethylzinc. The enantioenriched propargylic alcohols from this reaction serve as versatile synthetic intermediates and have enabled efficient syntheses of several complex natural products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50803
Propargyl alcohol, 99%
C3H4O