EMAIL THIS PAGE TO A FRIEND
DNA repair
DNA interstrand crosslinks induce a potent replication block followed by formation and repair of double strand breaks in intact mammalian cells.
PMID 23099010
Abstract
DNA interstrand crosslinks (ICLs) are highly toxic lesions that covalently link both strands of DNA and distort the DNA helix. Crosslinking agents have been shown to stall DNA replication and failure to repair ICL lesions before encountered by replication forks may induce severe DNA damage. Most knowledge of the ICL repair process has been revealed from studies in bacteria and cell extracts. However, for mammalian cells the process of ICL repair is still unclear and conflicting data exist. In this study we have explored the fate of psoralen-induced ICLs during replication, by employing intact mammalian cells and novel techniques. By comparative studies distinguishing between effects by monoadducts versus ICLs, we have been able to link the block of replication to the ICLs induction. We found that the replication fork was equally blocked by ICLs in wild-type cells as in cells deficient in ERCC1/XPF and XRCC3. The formation of ICL induced double strand breaks (DSBs), detected by formation of 53PB1 foci, was equally induced in the three cell lines suggesting that these proteins are involved at a later step of the repair process. Furthermore, we found that forks blocked by ICLs were neither bypassed, restarted nor restored for several hours. We propose that this process is different from that taking place following monoadduct induction by UV-light treatment where replication bypass is taking place as an early step. Altogether our findings suggest that restoration of an ICL blocked replication fork, likely initiated by a DSB occurs relatively rapidly at a stalled fork, is followed by restoration, which seems to be a rather slow process in intact mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0956
Angelicin
C11H6O3
P8399
Psoralen, ≥99% (HPLC)
C11H6O3
73633
Psoralen, analytical standard
C11H6O3