EMAIL THIS PAGE TO A FRIEND
Brain research
NGP1-01, a multi-targeted polycyclic cage amine, attenuates brain endothelial cell death in iron overload conditions.
PMID 23099055
Abstract
Development and progression of neurodegenerative disorders have, amongst other potential causes, been attributed to a disruption of iron regulatory mechanisms and iron accumulation. Excess extracellular iron may enter cells via nontraditional routes such as voltage-gated calcium channels and N-methyl-d-aspartate (NMDA) receptors leading to intracellular oxidative damage and ultimately mitochondrial failure. Nimodipine, an L-type calcium channel blocker has been shown to reduce iron-induced toxicity in neuronal and brain endothelial cells. Our current study investigates NGP1-01, a multimodal drug acting as an antagonist at both the NMDA receptor and the L-type calcium channel. Our previous studies support NGP1-01 as a promising neuroprotective agent in diseases involving calcium-related excitotoxicity. We demonstrate here that NGP1-01 (1 and 10μM) pretreatment abrogates the effects of iron overload in brain endothelial cells protecting cellular viability. Both concentrations of NGP1-01 were found to attenuate iron-induced reduction in cellular viability to a similar extent, and were statistically significant. To further verify the mechanism, the L-type calcium channel agonist FPL 64176 was administered to promote iron uptake. Addition of NGP1-01 dose-dependently reduced FPL 64176 stimulated uptake of iron. These data support further evaluation of NGP1-01 as a neuroprotective agent, not only in diseases associated with excitotoxicity, but also in those of iron overload.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F131
FPL 64176, ≥98% (HPLC), powder
C22H21NO3