EMAIL THIS PAGE TO A FRIEND
The American review of respiratory disease
A comparison of alpha 1-proteinase inhibitor methoxysuccinyl-Ala-Ala-Pro-Val-chloromethylketone and specific beta-lactam inhibitors in an acute model of human polymorphonuclear leukocyte elastase-induced lung hemorrhage in the hamster.
PMID 2310097
Abstract
A pharmacokinetic model is described for testing of polymorphonuclear leukocyte (PMN) elastase inhibitors administered by intratracheal or aerosol dosing of hamsters. Acute lung injury, measured as hemorrhage occurring within hours after intratracheal instillation of human PMN elastase, correlated directly with the amount of active enzyme instilled. Hemorrhage began within minutes of elastase instillation, was maximal within 1 h, and remained constant for up to 5 h subsequently. Therefore, inhibition of hemorrhage was used as an assay of the effectiveness of various PMN elastase inhibitors given by the intratracheal route. Lung hemorrhage could also be induced by intratracheal instillation of other elastolytic enzymes, such as thermolysin, and inhibition of hemorrhage was seen only with inhibitors active against the type of elastase used. Methoxysuccinyl-alanyl-alanyl-prolyl-valine-chloromethylketone (MeOSuc-AAPV-CMK), as well as alpha 1-proteinase inhibitor (alpha 1PI) but not tosyl-lysine-chloromethylketone (tosyl-lysine-CMK), inhibited the hemorrhage caused by human PMN elastase, but the specific inhibitors of this enzyme had no effect on thermolysin-induced lung hemorrhage. The duration of activity of these compounds as elastase inhibitors in this model correlated directly with the extent of their persistence in lung lavage fluid as determined by HPLC analysis of compound recovered by bronchoalveolar lavage. (ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0398
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val-chloromethyl ketone, elastase inhibitor
C22H35ClN4O7