EMAIL THIS PAGE TO A FRIEND
Harvard heart letter : from Harvard Medical School
Ask the doctors. My potassium level is high, and my physician stopped the medication that has successfully controlled my blood pressure for years, lisinopril. What is the danger from high potassium, and why is lisinopril a problem?
PMID 23101125
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6292
Lisinopril, ≥98% (HPLC)
C21H31O5N3 · 2H2O
PHR1143
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
C21H31O5N3 · 2H2O