EMAIL THIS PAGE TO A FRIEND
BMB reports
1,8-cineole protected human lipoproteins from modification by oxidation and glycation and exhibited serum lipid-lowering and anti-inflammatory activity in zebrafish.
PMID 23101510
Abstract
We recently reported that a water extract of laurel or turmeric, 1,8-cineole enriched fractions, showed hypolipidemic activity in the zebrafish model. Therefore, the present study investigated the cineole's anti-oxidant and anti-inflammatory activities in lipoprotein metabolism in vitro and in vivo. Cineole had inhibitory effects on cupric ion-mediated oxidation of lipoproteins in general, while simultaneously enhancing ferric ion removal ability in high-density lipoprotein (HDL). Hypercholesterolemia was induced in zebrafish using cholesterol-feeding treatment, 4% cholesterol, for 3 weeks. After feeding with or without the addition of cineole, the results revealed that cineole possessed lipid-lowering and anti-inflammatory activities in hypercholesterolemic zebrafish. In addition, serum amyloid A and interleukin-6 levels were lowered and lipid accumulation was decreased in the liver. Conclusively, 1,8-cineole was found to have anti-oxidant activities in lipoprotein metabolism both in vitro and in vivo with simultaneous reduction of lipid accumulation in the liver of zebrafish.