EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Antioxidant interactions between major phenolic compounds found in 'Ataulfo' mango pulp: chlorogenic, gallic, protocatechuic and vanillic acids.
PMID 23103532
Abstract
Phenolic compounds are known to have antioxidant capacity; however, there is little information about molecular interactions between particular phenolics found in fruits at different developmental stages. Therefore, the total antioxidant capacity of the phenolic compounds of a fruit may not correspond to the sum of individual antioxidant capacity given by antioxidants from that tissue. In this study, individual antioxidant capacity and the interactions of four major phenolic compounds (chlorogenic, gallic, protocatechuic and vanillic acid) found in 'Ataulfo' mango pulp were tested using the DPPH assay. Significant synergism was found in the majority of the all combinations, as well as the combination of the four phenolics. However, antagonism was also observed between some molecules. This work demonstrated particular interactions that may occur in a complex environment within the complex framework of a natural food. The present results may also assist in the future design of functional foods or ingredients based on their antioxidant activity and their synergistic or antagonist interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68654
Vanillic acid, certified reference material, TraceCERT®
C8H8O4