EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Development of a cancer-based chronic inhalation reference value for hexavalent chromium based on a nonlinear-threshold carcinogenic assessment.
PMID 23103602
Abstract
The carcinogenicity of hexavalent chromium(CrVI) is of significant interest to regulatory agencies for the protection of public health and to industry. Additionally, the mode of action (MOA) and conditions under which CrVI may induce carcinogenicity (e.g., reductive capacity considerations) have recently been the subject of significant scientific debate. Epidemiological data supported by data relevant to the carcinogenic MOA support considering nonlinear-threshold carcinogenic assessments for comparison to default linear low-dose extrapolation approaches. This study reviews epidemiological studies available in the scientific literature and conducts additional statistical dose-response analyses to identify potential carcinogenic thresholds and points of departure (PODs) in the context of supportive MOA information for a nonlinear-threshold inhalation carcinogenic assessment. Dosimetric adjustments and application of appropriate uncertainty factors (total UF of 30) to the selected cumulative exposure POD results in a cancer-based chronic inhalation reference value (ReV) of 0.24 μgCrVI/m(3). This chronic ReV is 300 times higher than the 1 in 100,000 excess cancer risk air concentration of 8E-04 μg/m(3) based on USEPA's unit risk factor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF81420094
Chromium, foil, 50x50mm, thickness 2.0mm, hard, 99.7+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF95051369
Chromium, foil, 25x25mm, thickness 2.0mm, hard, 99.7+%
Cr
GF15403047
Chromium, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF59666945
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF15970080
Chromium, foil, light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF50053092
Chromium, rod, 25mm, diameter 5.0mm, 99.7+%
Cr
GF22749240
Chromium, rod, 10mm, diameter 2.0mm, 99.7+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF31792956
Chromium, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr