EMAIL THIS PAGE TO A FRIEND
Journal of clinical orthodontics : JCO
Sterilization and instrument compatibility in a digital world.
PMID 23103851
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241881
Tungsten(IV) carbide, powder, 2 μm, ≥99%
CW