EMAIL THIS PAGE TO A FRIEND
Organic letters
Azobisisobutyronitrile initiated aerobic oxidative transformation of amines: coupling of primary amines and cyanation of tertiary amines.
PMID 23106189
Abstract
In the presence of a catalytic amount of radical initiator AIBN, primary amines are oxidatively coupled to imines and tertiary amines are cyanated to α-aminonitriles. These "metal-free" aerobic oxidative coupling reactions may find applications in a wide range of "green" oxidation chemistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
768375
Azobisisobutyronitrile, 12 wt. % in acetone
C8H12N4