EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Competitive inhibition of lipolytic enzymes. II. Preparation of 'monoacylamino' phospholipids.
PMID 2310761
Abstract
This paper describes the synthesis of a number of phosphatidylcholines and phosphatidylglycols, in which one fatty acyl ester group is replaced by an acylamino function. The phospholipids, both of the alpha- and beta-type, are prepared in racemic and enantiomeric pure forms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A76001
(±)-3-Amino-1,2-propanediol, 97%
C3H9NO2