EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
One-pot synthesis of responsive catalytic Au@PVP hybrid nanogels.
PMID 23108039
Abstract
Responsive catalytic hybrid nanogels with Au nanoparticle cores and a polyvinylpyrrolidone (PVP) based gel shell are prepared through a novel one-pot approach. The embedded Au nanoparticles demonstrate both a pH-modulated catalytic activity and anti-aggregation properties upon recycling.