EMAIL THIS PAGE TO A FRIEND
Arquivos brasileiros de cardiologia
Rosuvastatin and ciprofibrate in the treatment of dyslipidemia in patients with HIV.
PMID 23108642
Abstract
Dyslipidemia secondary to highly active antiretroviral therapy in patients with HIV is associated with a significant increase in cardiovascular morbidity and mortality due to atherosclerotic disease, requiring, thus, immediate and effective treatment. To demonstrate the effectiveness and safety of rosuvastatin and ciprofibrate in the treatment of dyslipidemia associated with highly active antiretroviral therapy in patients with HIV. Three hundred and forty-six patients with dyslipidemia underwent pharmacological treatment as follows: 200 patients with hypertriglyceridemia received ciprofibrate (Group I); 79 patients with hypercholesterolemia received rosuvastatin (Group II); and 67 patients with mixed dyslipidemia received ciprofibrate associated with rosuvastatin (Group III). The lipid profile was assessed before and after the lipid-lowering treatment, and the Wilcoxon test was used for statistical comparison. Liver transaminases and creatine phosphokinase were measured to assess liver and muscle toxicity. The serum concentrations of triglycerides and total cholesterol were significantly lower than those obtained before the lipid-lowering treatment in the three experimental groups (p < 0.002). A significant increase in HDL-cholesterol was observed in Groups I and III (p < 0.002). In Groups I and II, LDL-cholesterol was significantly lower (p < 0.001). None of the patients experienced elevations in transaminases or creatine phosphokinase to significantly toxic levels. The results of this study show that ciprofibrate and rosuvastatin or a combination of both can be considered an effective, safe and well-tolerated lipid-lowering treatment for patients with AIDS on highly active antiretroviral therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0330
Ciprofibrate
C13H14Cl2O3