EMAIL THIS PAGE TO A FRIEND
Molecular diversity
The synthesis of low molecular weight pyrrolo[2,3-c]pyridine-7-one scaffold.
PMID 23108947
Abstract
A facile method for the synthesis of substituted pyrrolo[2,3-c]pyridine-7-ones is developed that applies an acid-promoted intramolecular cyclization of 2-pyrrolecarboxylic acid amidoacetals as key step. The synthesis is easily scaled up to 1.5 mol quantity with no yield decrease. The alkylation/arylation reaction of the pyrrolo[2,3-c]pyridine-7-ones proceeds regioselectively giving N6-substituted derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P73609
Pyrrole-2-carboxylic acid, 99%
C5H5NO2