EMAIL THIS PAGE TO A FRIEND
Texas Heart Institute journal
Ergotamine-associated valvulopathy with recurrent chylous pleural effusion.
PMID 23109782
Abstract
We report a rare case of ergotamine-associated mitral stenosis in a 55-year-old woman who presented with recurrent chylous pleural effusion. Echocardiographic, gross, and microscopic features of the mitral valve were consistent with chronic ergotamine-induced valvulopathy. We conclude that medication-induced valvulopathy should be included in the differential diagnosis of valvular heart disease. In addition, cardiac function should be monitored before and during long-term therapy with ergotamine or ergotamine-derived dopamine agonists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45510
Ergotamine D-tartrate, ≥97.0% (calc. based on dry substance, NT)
C33H35N5O5 · 0.5C4H6O6