PloS one
NFAT1 C-terminal domains are necessary but not sufficient for inducing cell death.
PMID 23110116
The proteins belonging to the nuclear factor of activated T cells (NFAT) family of transcription factors are expressed in several cell types and regulate genes involved in differentiation, cell cycle and apoptosis. NFAT proteins share two conserved domains, the NFAT-homology region (NHR) and a DNA-binding domain (DBD). The N- and C-termini display two transactivation domains (TAD-N and TAD-C) that have low sequence similarity. Due to the high sequence conservation in the NHR and DBD, NFAT members have some overlapping roles in gene regulation. However, several studies have shown distinct roles for NFAT proteins in the regulation of cell death. The TAD-C shows low sequence similarity among NFAT family members, but its contribution to specific NFAT1-induced phenotypes is poorly understood. Here, we described at least two regions of NFAT1 TAD-C that confer pro-apoptotic activity to NFAT1. These regions extend from amino acids 699 to 734 and 819 to 850 of NFAT1. We also showed that the NFAT1 TAD-C is unable to induce apoptosis by itself and requires a functional DBD. Furthermore, we showed that when fused to NFAT1 TAD-C, NFAT2, which is associated with cell transformation, induces apoptosis in fibroblasts. Together, these results suggest that the NFAT1 TAD-C includes NFAT death domains that confer to different NFAT members the ability to induce apoptosis.
Related Materials