EMAIL THIS PAGE TO A FRIEND
Intensive care medicine
Beneficial effects of stress-dose corticosteroid therapy in canines depend on the severity of staphylococcal pneumonia.
PMID 23111805
Abstract
The effects of stress-dose corticosteroid therapy were studied in a canine staphylococcal pneumonia model of septic shock. Immediately following intrabronchial bacterial challenge, purpose-bred beagles were treated with stress doses of desoxycorticosterone (DOC), a mineralocorticoid agonist, and dexamethasone (DEX), a glucocorticoid agonist, or with placebo for 96 h. Oxacillin (30 mg/kg every 8 h) was started 4 h after infection onset. Bacterial dose was titrated to achieve 80-90 % lethality (n = 20) using an adaptive design; additional animals (n = 18) were investigated using the highest bacterial dose. Initial analysis of all animals (n = 38) demonstrated that the effects of DOC + DEX were significantly altered by bacterial dose (p = 0.04). The treatment effects of DOC + DEX were different in animals administered high or relatively lower bacterial doses in terms of survival (p = 0.05), shock reversal (p = 0.02), interleukin-6 levels (p = 0.02), and temperature (p = 0.01). DOC + DEX significantly improved the above parameters (p ≤ 0.03 for all) and lung injury scores (p = 0.02) after high-dose bacterial challenges, but not after lower challenges (p = not significant for all). Oxacillin trough levels were below the minimum inhibitory concentration of the infecting organism, and DOC + DEX increased the frequency of persistent staphylococcal bacteremia (odds ratio 3.09; 95 % confidence interval 1.05-9.11; p = 0.04). Stress-dose corticosteroids were only beneficial in cases of sepsis with high risk for death and even short courses may interfere with host mechanisms of bacterial clearance.
Related Materials