EMAIL THIS PAGE TO A FRIEND
Journal of physics. Condensed matter : an Institute of Physics journal
Depletion induced clustering in mixtures of colloidal spheres and fd-virus.
PMID 23114036
Abstract
We determined the phase boundary of an ideal rod-sphere mixture consisting of fd-virus, which is an established model system for mono-disperse colloidal rods, and density matched mono-disperse polystyrene beads employing diffuse wave spectroscopy. The low volume fraction of fd needed to induce a phase separation at relatively low ionic strength exemplifies the fact that slender rods are very effective depletion agents. Confocal microscopy showed that stable clusters are formed during phase separation. Relaxation after shear deformation of these clusters showed that the phase separation is gas-liquid-like and that the interfacial tension involved is very low as in colloid-polymer mixtures.